Skip to Content
Merck
All Photos(1)

Key Documents

GF326

Sigma-Aldrich

IL-13,
Human Recombinant Animal Free

Synonym(s):

Interleukin-13, IL13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77
Pricing and availability is not currently available.

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

General description

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and also by mast cells and NK cells. Targeted deletion of IL-13 in mice resulted in impaired Th2 cell development and indicated an important role for IL-13 in the expulsion of gastrointestinal parasites. IL-13 exerts anti-inflammatory effects on monocytes and macrophages and it inhibits the expression of inflammatory cytokines such as IL-1β, TNF-α, IL-6 and IL-8. IL-13 has also been shown to enhance B cell proliferation and to induce isotype switching resulting in increased production of IgE. Blocking of IL-13 activity inhibits the pathophysiology of asthma. Human and murine IL-13 is cross-species reactive.
Recombinant human IL-13 is a 12.6 kDa protein consisting of 115 amino acid residues.
Product Source: Protein is expressed in E.coli.
The protein is manufactured without the use of any animal products/reagents and is considered Animal-Free and can be used in protocols where possible transmisson of animal viruses and contamination must be avoided.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

The ED50 was determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥1 x 10^6
units/mg.

Physical form

Product is presented in 20 mM Sodium Phosphate, pH 7.0 and is filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt Centrifuge the vial prior to opening.
Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein and store in working aliquots at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service