BI0420
Oxidizing Reagent, .02M I₂O in THF/Pyridine/H2O (70/20/10)
Biosynthesis Novabiochem®
Synonym(s):
DNA Reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116006
Recommended Products
Quality Level
product line
Novabiochem®
form
liquid
reaction suitability
reaction type: Oxidations
reagent type: oxidant
transition temp
flash point -13 °C
shipped in
ambient
storage temp.
2-30°C
Application
BI0420-0201 is packaged in 24/400 glass bottles
BI0420-0200 is packaged in 28/405 glass bottles
BI0420-0450 is packaged in 28/405 glass bottles
BI0420-0960 is packaged in 28/405 glass bottles
BI0420-P4000 is packaged in poly coated glass bottles
BI0420-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
BI0420-0200 is packaged in 28/405 glass bottles
BI0420-0450 is packaged in 28/405 glass bottles
BI0420-0960 is packaged in 28/405 glass bottles
BI0420-P4000 is packaged in poly coated glass bottles
BI0420-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®
Analysis Note
Color: Red
Form: Liquid
Iodine Molarity: 0.0190-0.0210M
Water (H2O): 9.0-11.0%
Form: Liquid
Iodine Molarity: 0.0190-0.0210M
Water (H2O): 9.0-11.0%
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
8.6 °F
Flash Point(C)
-13 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service