Skip to Content
Merck
All Photos(3)

Key Documents

AB5210

Sigma-Aldrich

Anti-Sodium Channel Antibody, Voltage Gated, Pan (SP19 Segment)

Chemicon®, from rabbit

Synonym(s):

Anti-DEE6, Anti-DEE6B, Anti-DRVT, Anti-EIEE6, Anti-FEB3A, Anti-FHM3, Anti-GEFSP2, Anti-HBSCI, Anti-NAC1, Anti-Nav1.1, Anti-SCN1, Anti-SMEI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SCN1A(6323)

Specificity

Recognizes alpha subunits of VGSC. Does not cross react with any other sodium channel antigens tested so far.

Immunogen

Purified peptide corresponding to amino acids 1501-1518 in the alpha subunit (amino acids 1501-1518) of rat type I voltage-gated sodium channel (VGSC) (Accession P04774).

Application

All procedures that are going to receive a full-length protein should be performed at 4°C, and the following protease inhibitor mixture should be used: pepstatin A (1 μg/mL), leupeptin (1 μg/mL), aprotinin (1 μg/mL), Pefabloc SC (0.2 mM), benzamidine (0.1 mg/mL), and calpain inhibitors I and II (8 μg/mL each).

Western blot: 1:200 using ECL on newborn rat brain membranes.

Immunohistochemistry on mouse brain sections (frozen) and mouse spinal cord sections (paraffin).

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Neuronal & Glial Markers
This Sodium Channel antibody is validated for use in IHC & WB for the detection of the Sodium Channel protein.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 200 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen. The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yumi Matsushita et al.
Biomedical research (Tokyo, Japan), 39(4), 189-196 (2018-08-14)
Ten pairs of protrusions, called accessory lobes (ALs), exist at the lateral sides of the avian lumbosacral spinal cord. Histological evidence indicates that neuron-like cells gather in the ALs, and behavioral evidence suggests that the ALs act as a sensory
Elsa Cisneros et al.
Frontiers in cellular neuroscience, 9, 470-470 (2015-12-24)
Kv7.2 channel expression has been reported to decrease in dorsal root ganglia (DRG) following the induction of a peripheral neuropathy while other experiments show that Kv7.2 accumulates in peripheral neuromas. The mechanisms underlying these novel expression patterns are poorly understood.
John E Rash et al.
Journal of neurophysiology, 115(4), 1836-1859 (2016-01-15)
Saltatory conduction in mammalian myelinated axons was thought to be well understood before recent discoveries revealed unexpected subcellular distributions and molecular identities of the K(+)-conductance pathways that provide for rapid axonal repolarization. In this study, we visualize, identify, localize, quantify
Claudia Kuehn et al.
The European journal of neuroscience, 37(6), 860-875 (2013-01-03)
Input-output computations of individual neurons may be affected by the three-dimensional structure of their dendrites and by the location of input synapses on specific parts of their dendrites. However, only a few examples exist of dendritic architecture which can be
Tristan D Kubik et al.
Scientific reports, 11(1), 7301-7301 (2021-04-02)
Aedes aegypti is a major vector of Zika, dengue, and other arboviruses. Permethrin adulticidal spraying, which targets the voltage-gated sodium channel (VGSC), is commonly done to reduce local mosquito populations and protect humans from exposure to arbovirus pathogens transmitted by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service