Skip to Content
Merck
All Photos(1)

Key Documents

8.53034

Sigma-Aldrich

Boc-His(Trt)-OH

≥98.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Boc-His(Trt)-OH, N-α-t.-Boc-N-im-trityl-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H31N3O4
CAS Number:
Molecular Weight:
497.58
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Product Name

Boc-His(Trt)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥95.0% (acidimetric)
≥98% (TLC)
≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

130 °C (decomposition)

application(s)

peptide synthesis

storage temp.

15-25°C

SMILES string

[n]4(cnc(c4)C[C@H](NC(=O)OC(C)(C)C)C(=O)O)C(c3ccccc3)(c2ccccc2)c1ccccc1

InChI

1S/C30H31N3O4/c1-29(2,3)37-28(36)32-26(27(34)35)19-25-20-33(21-31-25)30(22-13-7-4-8-14-22,23-15-9-5-10-16-23)24-17-11-6-12-18-24/h4-18,20-21,26H,19H2,1-3H3,(H,32,36)(H,34,35)/t26-/m0/s1

InChI key

OYXZPXVCRAAKCM-SANMLTNESA-N

General description

A protected histidine derivative for only the critical coupling step in Boc SPPS. After coupling the Boc- and Trt-groups are cleaved simultaneously by TFA.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Linkage

Replaces: 04-12-0160

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 95.0 %
Water (K. F.): ≤ 4.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service