Skip to Content
Merck
All Photos(1)

Key Documents

8.40041

Sigma-Aldrich

2-Phenylpyridine

for synthesis

Synonym(s):

2-Phenylpyridine, 2-Azabiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N
CAS Number:
Molecular Weight:
155.20
MDL number:
UNSPSC Code:
12352005
EC Index Number:
213-763-1
NACRES:
NA.22

vapor pressure

<0.5 hPa ( 30 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

269-270 °C/1013 hPa

transition temp

flash point 110 °C

density

1.09 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

n1c(cccc1)c2ccccc2

InChI

1S/C11H9N/c1-2-6-10(7-3-1)11-8-4-5-9-12-11/h1-9H

InChI key

VQGHOUODWALEFC-UHFFFAOYSA-N

Application

2-Phenylpyridine for synthesis. CAS 1008-89-5, molar mass 155.2 g/mol.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.086 - 1.090
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service