Skip to Content
Merck
All Photos(1)

Key Documents

8.20956

Sigma-Aldrich

2,2,3,3,3-Pentafluoropropionic acid

for synthesis

Synonym(s):

2,2,3,3,3-Pentafluoropropionic acid, Perfluoropropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3HF5O2
CAS Number:
Molecular Weight:
164.03
MDL number:
UNSPSC Code:
12352106
EC Index Number:
207-021-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

52 hPa ( 25 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

bp

96-97 °C/984 hPa

density

1.57 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Ag]OC(=O)C(F)(F)C(F)(F)F

InChI

1S/C3HF5O2.Ag/c4-2(5,1(9)10)3(6,7)8;/h(H,9,10);/q;+1/p-1

InChI key

XAYJXAUUXJTOSI-UHFFFAOYSA-M

Related Categories

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.571 - 1.575
Colour (visual): colorless to yellowish to pink to brownish
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service