8.14325
Silver trifluoromethanesulfonate
for synthesis
Synonym(s):
Silver trifluoromethanesulfonate, Trifluoromethanesulfonic acid silver salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CAgF3O3S
CAS Number:
Molecular Weight:
256.94
MDL number:
UNSPSC Code:
12352103
EC Index Number:
220-882-2
NACRES:
NA.22
Recommended Products
Quality Level
form
powder
mp
356 °C
storage temp.
2-30°C
SMILES string
[Ag+].FC(F)(F)[S](=O)(=O)[O-]
InChI
1S/CHF3O3S.Ag/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1
InChI key
QRUBYZBWAOOHSV-UHFFFAOYSA-M
Analysis Note
Assay (argentometric): ≥ 97.0 %
Identity (IR): passes test
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service