Skip to Content
Merck
All Photos(2)

Key Documents

8.03237

Sigma-Aldrich

2-(Dimethylamino)-ethanol

for synthesis

Synonym(s):

2-(Dimethylamino)-ethanol, N,N-Dimethylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2OH
CAS Number:
Molecular Weight:
89.14
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-542-8
NACRES:
NA.22

vapor pressure

6.12 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

245 °C

potency

2130 mg/kg LD50, oral (Rat)
1220 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-12.2 % (v/v)

pH

10.5-11 (20 °C, 100 g/L in H2O)

kinematic viscosity

3.58 cSt(21.6 °C)

mp

-59 °C

transition temp

flash point 40 °C (DIN 51755 Part 1)

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3

InChI key

UEEJHVSXFDXPFK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.886 - 0.887
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - Seta closed cup

Flash Point(C)

40 °C - Seta closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service