Skip to Content
Merck
All Photos(1)

Key Documents

8.01587

Sigma-Aldrich

1-Butanethiol

for synthesis

Synonym(s):

1-Butanethiol, Butyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SH
CAS Number:
Molecular Weight:
90.19
MDL number:
UNSPSC Code:
12352105
EC Index Number:
203-705-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

61 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

1500 mg/kg LD50, oral (Rat)

expl. lim.

1.4-11.3 % (v/v)

bp

97-99 °C/1013 hPa

mp

-116 °C

transition temp

flash point 12 °C

solubility

0.597 g/L

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCCCC

InChI

1S/C4H10S/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

WQAQPCDUOCURKW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.840 - 0.842
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service