Skip to Content
Merck
All Photos(1)

Key Documents

71110-M

Millipore

rLysozyme Solution

Stabilized recombinant lysozyme

Synonym(s):

recombinant lysozyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

form

solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

rLysozyme Solution contains a highlypurified and stabilized recombinant lysozymethat can be used for lysis of Gram-negativebacteria, such as E. coli. The enzyme catalyzes the hydrolysis of N-acetylmuramide linkages in bacterial cell walls. The specific activity of rLysozyme (1700 KU/mg) for E. coli lysis is 250 times greater than that of chicken egg white lysozyme. rLysozyme is optimally active at physiological pH. Very small amounts of rLysozyme (3–5 KU/g cell paste) enhance the efficiency of protein extraction withBugBuster, BugBuster HT, and PopCultureReagents. In the absence of protein extractionreagents, direct lysis of E. coli can be achieved by treatment of 1.0 gram cell paste with 45–60 KU rLysozyme. The product is supplied as a ready-to-use solution at a concentration range of 27–33 KU/µl in 50% glycerol containing 50 mM Tris-HCl, 0.1 M NaCl, 0.1 mM EDTA, 1 mM DTT, 0.1% TRITON X-100, pH 7.5. rLysozyme Solution is stable at –20°C.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit of rLysozyme is defined as the amount of enzyme necessary to cause a decrease of 0.025 A₄₅₀ unit per min at 25°C in a 1-ml suspension (1 mg/ml) of Tuner(DE3) cells in 0.5 x BugBuster diluted with 50 mM Tris-HCl, pH 7.5.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cell lysis and nucleic acid removal for Gram-negative and Gram-positive bacteria using the BugBuster Plus Lysonase™ Kit

The combination of BugBuster® and Lysonase™ reagents greatly enhances the release of soluble proteins from Gram-positive bacteria.

Related Content

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service