Skip to Content
Merck
All Photos(2)

Key Documents

666122-M

Millipore

Urea

Molecular Biology Grade, End product of the urea cycle. A topical keratolytic agent.

Synonym(s):

Urea, Carbamide, Carbonyl Diamide, Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.25
grade:
Molecular Biology

grade

Molecular Biology

Quality Level

description

Merck USA index - 14, 9867

Assay

≥98%

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

132-135 °C (lit.)

solubility

water: soluble

density

1.335 g/mL at 25 °C (lit.)

absorption

≤0.01 at 260 nm
≤0.01 at 280 nm

foreign activity

DNases, proteases, RNases, none detected

storage temp.

10-30°C

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

End product of the urea cycle (absorbance: ≤0.01 at 260 nm and 280 nm). A topical keratolytic agent.
End product of the urea cycle. A topical keratolytic agent.

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service