561505
L-Selenomethionine
L Selenomethionine increases the expression of glutathione peroxidase.
Synonym(s):
L-Selenomethionine, SeMet
About This Item
Recommended Products
Quality Level
description
Merck USA index - 14, 8441
Assay
≥97% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white to off-white
solubility
water: 50 mg/mL
cation traces
Se: ≥39.5%
heavy metals: ≤10 ppm
shipped in
ambient
storage temp.
15-25°C
SMILES string
[Se](CC[C@H]([N+H3])C(=O)[O-])C
InChI
1S/C5H11NO2Se/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1
InChI key
RJFAYQIBOAGBLC-BYPYZUCNSA-N
General description
Warning
Reconstitution
Other Notes
Boles, J.O., et al. 1992. J. Biol. Chem.267, 22217.
Nielson, J.B. and Anderson, O. 1992. Toxicology74, 233.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
250.3 - 346.3 °F
Flash Point(C)
121.3 - 174.6 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service