5.09612
TAPI-2
InSolution, ≥97%, 10 mM aqueous solution, inhibitor of matrix metalloproteinases and TACE (TNF-α convertase; ADAM17)
Synonym(s):
InSolution TAPI-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H37N5O5
CAS Number:
Molecular Weight:
415.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
storage temp.
−70°C
General description
A hydroxamate-based inhibitor of matrix metalloproteinases and TACE (TNF-α convertase; ADAM17). Inhibits the activation-dependent shedding of L-selectin from neutrophils, eosinophils, and lymphocytes as well as phenylarsine oxide (PAO)-induced shedding in neutrophils. Also blocks the shedding of several cell surface proteins such as pro-TGF-α, IL-6 receptor α-subunit, and β-amyloid precursor protein. Reported to inhibit TNF-α release and impart protection against aspirin-induced gastric mucosal injury. Acts as a weak inhibitor of angiotensin converting enzyme (IC50 = 18 µM).
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Physical form
A 10 mM (1 mg/241 µL) sterile-filtered solution of TAPI-2 (Cat. No. 579052) in H₂O.
Reconstitution
Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 3 months at -20°C.
Other Notes
Bennett, T.A., et al. 2000. J. Immunol.164, 4120.
Fiorucci, S., et al. 1998. Aliment. Pharmacol. Ther.12, 1139.
Hooper, N.M., et al. 1997. Biochem. J.321, 265.
Parvathy, S., et al. 1997. Biochem. J.327, 37.
Arribas, J., et al. 1996. J. Biol. Chem.271, 11376.
Fiorucci, S., et al. 1998. Aliment. Pharmacol. Ther.12, 1139.
Hooper, N.M., et al. 1997. Biochem. J.321, 265.
Parvathy, S., et al. 1997. Biochem. J.327, 37.
Arribas, J., et al. 1996. J. Biol. Chem.271, 11376.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service