Skip to Content
Merck
All Photos(1)

Key Documents

5.04297

Sigma-Aldrich

RIP1 Inhibitor II, 7-Cl-O-Nec-1

Synonym(s):

RIP1 Inhibitor II, 7-Cl-O-Nec-1, 7-Cl-O-MH-Trp, Nec-1s, Nec-1 stable, RIPK1 Inhibitor II, Receptor-Interacting Protein 1 Inhibitor II, Necrosis Inhibitor IV, 5-((7-Chloro-1H-indol-3-yl)methyl)-3-methyl-2,4-imidazolidinedione, 5-((7-Chloro-1H-indol-3-yl)methyl)-3-methylimidazolidine-2,4-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12ClN3O2
CAS Number:
Molecular Weight:
277.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to pale yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

Clc1c2[nH]cc(c2ccc1)CC3NC(=O)N(C3=O)C

InChI

1S/C13H12ClN3O2/c1-17-12(18)10(16-13(17)19)5-7-6-15-11-8(7)3-2-4-9(11)14/h2-4,6,10,15H,5H2,1H3,(H,16,19)

InChI key

WIKGAEMMNQTUGL-UHFFFAOYSA-N

General description

A cell-permeable 7-chloroindolylmethyl-hydantoin that exhibits highly RIP1-selective inhibitory activity among >400 human kinases and is superior to its structural analog Necrostatin-1 (Cat. No. 480065) in blocking RIP1-dependent necroptosis due to improved potency (EC50 = 206 nM vs. 494 nM with Nec-1 for protecting FADD-deficient Jurkat from 30 h 10 ng/mL TNF-α-induced death), selectivity, metabolic stability, as well as reduced in vitro and in vivo toxicity. Reported to be blood-brain barrier permeable in mice and be efficacious in reducing brain infarct size when applied via intracerebroventricular injections in a murine MCAO model in vivo (8 nmol/2 µL/mouse).
A cell-permeable 7-chloroindolylmethyl-hydantoin that exhibits highly RIP1-selective inhibitory activity among >400 human kinases and is superior to its structural analog Necrostatin-1 (Cat. No. 480065) in blocking RIP1-dependent necroptosis due to improved potency (EC50 = 206 nM vs. 494 nM with Nec-1 for protecting FADD-deficient Jurkat from 30 h 10 ng/mL TNF-α-induced death), selectivity (no inhibition against IDO/indolamine 2,3-deoxygenase vs. IC50 = 11.4 µM with Nec-1), metabolic stability, as well as reduced in vitro and in vivo toxicity. In addition, 7-Cl-O-Nec-1 is reported to exhibit blood-brain barrier permeability in mice (Brain and plasma conc. = 0.74 and 0.31 µM, respectively, 30 min post 1 mg/6.25 mL/kg i.v. injection using male mice) and is efficacious in reducing brain infarct size when applied via intracerebroventricular injections (63% of control size 18 h post occlusion; two 8 nmol/2 µL/mouse injections at 4 h & 6 h post occlusion) in a murine MCAO model in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RIP1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Ofengeim, D., et al. 2015. Cell Reports10, In press.
Muller, A.J., et al. 2005. Nat. Med.11, 312.
Degterev, A., et al. 2013. Nat. Chem. Biol.9, 192.
Degterev, A., et al. 2013. Cell Death Differ.20, 366.
Christofferson, D.E., et al. 2012. Cell Death Dis.3, e320.
Takahashi, N., et al. 2012. Cell Death Dis.3, e437.
Degterev, A., et al. 2008. Nat. Chem. Biol.4, 313.
Degterev, A., et al. 2005. Nat. Chem. Biol.1, 112.
Teng, X., et al. 2005. Bioorg. Med. Chem. Lett.15, 5039.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Wang et al.
Frontiers in cellular and infection microbiology, 9, 286-286 (2019-08-24)
Programmed cell death and especially necroptosis, a programmed and regulated form of necrosis, have been recently implicated in the progression and outcomes of influenza in mouse models. Moreover, Z-DNA/RNA binding protein 1 (ZBP1) has been identified as a key signaling
Andre L Samson et al.
Nature communications, 11(1), 3151-3151 (2020-06-21)
Mixed lineage kinase domain-like (MLKL) is the terminal protein in the pro-inflammatory necroptotic cell death program. RIPK3-mediated phosphorylation is thought to initiate MLKL oligomerization, membrane translocation and membrane disruption, although the precise choreography of events is incompletely understood. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service