Skip to Content
Merck
All Photos(1)

Documents

499609-M

Millipore

(5Z)-Zeaenol, Curvularia sp.

A cell-permeable resorcylic acid lactone compound that may serve as a negative control for TAK1 (TGF-β-Activated Kinase-1) inhibition studies.

Synonym(s):

(5Z)-Zeaenol, Curvularia sp., TAK1 Inhibitor, Inactive Control

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O7
CAS Number:
Molecular Weight:
364.39
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

storage temp.

−20°C

InChI

1S/C19H24O7/c1-11-5-3-7-14(20)18(23)15(21)8-4-6-12-9-13(25-2)10-16(22)17(12)19(24)26-11/h3-4,6-7,9-11,14-15,18,20-23H,5,8H2,1-2H3

InChI key

BPOLRDGTYHVUAY-UHFFFAOYSA-N

General description

A cell-permeable resorcylic acid lactone compound that may serve as a negative control for TAK1 (TGF-β-Activated Kinase-1) inhibition studies. The active TAK1 inhibitor is also available, (5Z)-7-Oxozeaenol, Curvularia sp. (Cat. No. 499610).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ninomiya-Tsuji, J., et al. 2003. J. Biol. Chem.278, 18485.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service