Skip to Content
Merck
All Photos(1)

Key Documents

495414

Sigma-Aldrich

1-Oleoyl-2-acetyl-sn-glycerol

Cell-permeable diacylglycerol analog that activates Ca2+-dependent protein kinase C.

Synonym(s):

1-Oleoyl-2-acetyl-sn-glycerol, OAG, PKC Activator III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H42O5
CAS Number:
Molecular Weight:
398.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: soluble
ethanol: soluble

format

neat

shipped in

ambient

storage temp.

−20°C

SMILES string

O([C@H](COC(=O)CCCCCCC\C=C/CCCCCCCC)CO)C(=O)C

InChI

1S/C23H42O5/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(26)27-20-22(19-24)28-21(2)25/h10-11,22,24H,3-9,12-20H2,1-2H3/b11-10-/t22-/m0/s1

InChI key

PWTCCMJTPHCGMS-YRBAHSOBSA-N

General description

Cell-permeable diacylglycerol analog that activates Ca2+-dependent protein kinase C. Also a stimulator of secretion and aggregation in blood platelets.
Note: 1 set = 10 x 500 µg.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Ca2+-dependent protein kinase C
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use. The material can be dissolved in chloroform, aliquoted, dried under a stream of nitrogen, and stored at -20°C.

Other Notes

Florin-Christensen, J. 1993. Biochem. J.289, 783.
Benz, I., et al. 1992. J. Membr. Biol. 130, 183.
Haeffner, E.W., and Wittmann, U. 1992. J. Lipid Mediat.5, 237.
Gilmore, T., and Martin, G.S. 1983. Nature306, 487.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service