Skip to Content
Merck
All Photos(1)

Key Documents

475874-M

Millipore

MK571, Sodium Salt

A selective, competitive antagonist of leukotriene D4 (LTD4) (Ki = 2.1 nM for inhibition of [³H]LTD4 binding to human lung membranes).

Synonym(s):

MK571, Sodium Salt, L 660711, Sodium Salt, (E)-3-[[[3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-[[3-dimethylamino)-3-oxopropyl]thio]methyl]thio]-propanoic Acid, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26ClN2O3S2 · Na
CAS Number:
Molecular Weight:
537.07
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

DMSO: 10 mg/mL
water: 200 mg/mL

storage temp.

−20°C

SMILES string

S(C(SCCC(=O)O)c1cc(ccc1)\C=C\c2nc3c(cc2)ccc(c3)Cl)CCC(=O)N(C)C

InChI

1S/C26H27ClN2O3S2/c1-29(2)24(30)12-14-33-26(34-15-13-25(31)32)20-5-3-4-18(16-20)6-10-22-11-8-19-7-9-21(27)17-23(19)28-22/h3-11,16-17,26H,12-15H2,1-2H3,(H,31,32)/b10-6+

InChI key

AXUZQJFHDNNPFG-UXBLZVDNSA-N

General description

A selective, competitive antagonist of leukotriene D4 (LTD4) receptor (Ki = 2.1 nM for inhibition of [3H]LTD4 binding to human lung membranes). Also reported to inhibit multidrug resistance-associated protein 1 (MRP1).

Biochem/physiol Actions

Primary Target
Antagonist of leukotriene D4 (LTD4)
Target Ki: 2.1 nM for inhibition of [3H]LTD4 binding to human lung membranes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions in are stable for up to 1 week at -20°C; DMSO stock solutions are stable for up to 6 months at -20°C. Acidic pH and heavy metals will accelerate decomposition.

Other Notes

Olson, D.P., et al. 2001. Cytometry46, 105.
Van der Kolk, D.M., et al. 1998. Clin. Cancer Res.4, 1727.
Jones, T.R., et al. 1989. Can. J. Physiol. Pharmacol.67, 17.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gad D Vatine et al.
Cell stem cell, 20(6), 831-843 (2017-05-21)
Inactivating mutations in the thyroid hormone (TH) transporter Monocarboxylate transporter 8 (MCT8) cause severe psychomotor retardation in children. Animal models do not reflect the biology of the human disease. Using patient-specific induced pluripotent stem cells (iPSCs), we generated MCT8-deficient neural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service