Skip to Content
Merck
All Photos(2)

Key Documents

373227

Sigma-Aldrich

MDM2 Inhibitor VII, MEL23

The MDM2 Inhibitor VII controls the biological activity of MDM2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MDM2 Inhibitor VII, MEL23, MDM2-MDMX E3 Ligase Inhibitor, 3-Butyl-6-hydroxy-5-(2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indol-1-yl)pyrimidine-2,4(1H,3H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N4O3
CAS Number:
Molecular Weight:
354.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 2.5 mg/mL, clear, pale yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC1=C(C(N(CCCC)C(N1)=O)=O)C(NCC2)C3=C2C4=C(N3)C=CC=C4

General description

A cell-permeable tetrahydro-β-carbolinylbarbiturate compound that selectively inhibits the E3 ligase activity of Mdm2-MdmX hetero-complex over that of Mdm2-Mdm2 homo-complex (70.6% vs. 17.6% inhibition, respectively, at 100 µM), without affecting Mdm2-MdmX complex formation or the activity of two other UbcH5C-utilizing ligase complexes Roc1-Cul1 and BRCA1-BARD1. Effectively inhibits ubiquitination and proteasomal degradation of cellular Mdm2 and p53 (effective conc. = 14 µM in U2OS, RKO, and HCT116 cultures) and induce RKO and MEF cell death in a p53- and Mdm2-dependent manner. Unlike Nutlin-3 (Cat. Nos. 444143 & 444151), MEL23 does not interfere with Mdm2-p53 interaction.
A cell-permeable tetrahydro-β-carbolinylbarbiturate compound that selectively inhibits the E3 ligase activity of Mdm2-MdmX hetero-complex over that of Mdm2-Mdm2 homo-complex (70.6% vs. 17.6% inhibition, respectively, with 100 µM inhibitor), without affecting Mdm2-MdmX complex formation or the activity of Roc1-Cul1 and BRCA1-BARD1, two other ligase complexes that also utilize UbcH5C as the Ub-conjugating enzyme. Effectively inhibits Mdm2-MdmX E3 ligase activity-dependent ubiquitination and proteasomal degradation of cellular Mdm2 and p53 (effective conc. = 14 µM in U2OS, RKO, and HCT116 cultures) and prevents DNA damage-induced MdmX degradation in MCF7 cells upon 50 µM etoposide treatment. Shown to induce RKO and MEF cell death in a p53- and Mdm2-dependent manner. Unlike Nutlin-3 (Cat. Nos. 444143 & 444151), MEL23 does not act by interfering Mdm2-p53 interaction.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Herman, A.G., et al. 2011. Cancer Discovery1, 312.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alyssa M Klein et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(44) (2021-10-31)
The p53 tumor suppressor protein, known to be critically important in several processes including cell-cycle arrest and apoptosis, is highly regulated by multiple mechanisms, most certifiably the Murine Double Minute 2-Murine Double Minute X (MDM2-MDMX) heterodimer. The role of MDM2-MDMX
Rafaela Muniz de Queiroz et al.
Nature communications, 15(1), 7132-7132 (2024-08-21)
Although the E3 ligase Mdm2 and its homologue and binding partner MdmX are the major regulators of the p53 tumor suppressor protein, it is now evident that Mdm2 and MdmX have multiple functions that do not involve p53. As one

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service