Skip to Content
Merck
All Photos(1)

Key Documents

324385

Sigma-Aldrich

Doxycycline, Hyclate

A commonly-used broad-spectrum antibiotic.

Synonym(s):

Doxycycline, Hyclate, α-6-Deoxy-5-hydroxytetracycline, Vibramycin, Hyclate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · 0.5H2O · 0.5C2H6O · xHCl
CAS Number:
Molecular Weight:
476.48 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

yellow to yellow-green

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/2C22H24N2O8.C2H6O.2ClH.H2O/c2*1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;1-2-3;;;/h2*4-7,10,14-15,17,25-27,31-32H,23H2,1-3H3;3H,2H2,1H3;2*1H;1H2/b2*21-13+;;;;/t2*7-,10+,14+,15-,17-,22-;;;;/m00..../s1

InChI key

QDDHZFKCHWYXJF-PCLMCNLLSA-N

General description

A commonly used broad-spectrum antibiotic. A potent inhibitor of MMP-8 (neutrophil collagenase; IC50 = 30 µM) relative to MMP-1 (collagenase-1; IC50 = 300 µM). Also inhibits collagen synthesis by bovine articular chondrocytes cultured in alginate (IC50 = 25 µM). Inhibits nitric oxide synthesis by LPS-activated macrophages without affecting IL-10 release.
A commonly-used broad-spectrum antibiotic. A potent inhibitor of MMP-8 (neutrophil collagenase; IC50 = 30 µM) relative to MMP-1 (interstitial collagenase; IC50 = 300 µM). Also inhibits collagen synthesis by bovine articular chondrocytes cultured in alginate (IC50 = 25 µM). Inhibits nitric oxide synthesis by LPS-activated macrophages without affecting IL-10 release.

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMP-8
Product does not compete with ATP.
Reversible: no
Target IC50: 30 µM against MMP-8 (neutrophil collagenase); 25 µM inhibiting collagen synthesis by bovine articular chondrocytes cultured in alginate

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Follwing reconstitution, refrigerate (4°C). Stock solutions are stable for up to 5 days at 4°C.

Other Notes

D’Agostino, P., et al. 1998. J. Infect. Dis. 177, 489.
Beekman, B., et al. 1997. Biochem. Biophys. Res. Commun. 237, 107.
Hanemaaijer, R., et al. 1997. J. Biol. Chem. 272, 31504.
Lauhio, A., et al. 1994. Clin. Exp. Immunol. Antimicrobiol. Agents Chemother. 36, 227.
Sorsa, T., et al. 1994. Ann. N.Y. Acad. Sci.732, 112.
Suomalainen, K., et al. 1992. Antimicrobiol. Agents Chemother.36, 227.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan C Shepherd et al.
Cell communication and signaling : CCS, 19(1), 100-100 (2021-10-09)
Fetal cell-derived exosomes (extracellular vesicles, 40-160 nm) are communication channels that can signal parturition by inducing inflammatory changes in maternal decidua and myometrium. Little is known about maternal cell-derived exosomes and their functional roles on the fetal side. This study isolated
Hsiang-Chen Chou et al.
eLife, 10 (2021-02-02)
The origin recognition complex (ORC) cooperates with CDC6, MCM2-7, and CDT1 to form pre-RC complexes at origins of DNA replication. Here, using tiling-sgRNA CRISPR screens, we report that each subunit of ORC and CDC6 is essential in human cells. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service