Skip to Content
Merck
All Photos(2)

Key Documents

1.46539

Millipore

Tryptic Soy Agar - Ready-to-use Contact Plates

Cephase, Lecithin, Tween®, ICR plus with lockable lid, plate diam. 55 mm, sterile; γ-irradiated, pkg of 10 plates triple packed, suitable for surface monitoring

Synonym(s):

Soybean Casein Digest Agar with neutralizers and Cephase, TSA, TSA + LT + Cephase, Tryptic Soy Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106231
NACRES:
NA.74
Pricing and availability is not currently available.

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate (ready-to-use)

feature

ready-to-use

composition

Cephase
Lecithin
Tween®

packaging

pkg of 10 plates triple packed

plate diam.

55 mm

pH

7.3 ( in H2O)

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

storage temp.

2-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Contact Agar with Cephase + LT – ICR+ is designed for the determination of the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Clean rooms and Isolators. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. The medium contains Cephase to inactivate a broad range of beta lactam-antibiotics and is supplemented with the two neutralizers lecithin and polysorbate (Tween®) 80, to inactivate possible residues of disinfectants on dry surfaces. In addition, the ICRplus (plus or “+”) plates are supplied with a lockable lid for safe transport after sampling without the risk of losing the lid. ICRplus plates are suitable for aerobic and anaerobic incubation by turning the lid into the close position for aerobic incubation and into the vent position for anaerobic incubation

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic count on dry, sanitized surfaces and personnel in Isolators and Clean Rooms.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service