Skip to Content
Merck
All Photos(1)

Key Documents

1.46266

Millipore

ReadyPlate CT DG 18 Agar ISO 21527-2

ReadyPlate, ready-to-use, plate diam. 55 mm, for yeasts, for molds

Synonym(s):

DG 18 Agar, Dichloran 18% Glycerol Chloramphenicol Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

Agency

ISO 21527-2

Quality Level

sterility

sterile

product line

ReadyPlate

form

plate (ready-to-use)

feature

ready-to-use

composition

18% (w/w) glycerol and chloramphenicol

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate CT

plate diam.

55 mm

pH

5.6±0.2

application(s)

food and beverages
surface monitoring

storage temp.

15-25°C

suitability

molds
yeasts

General description

ReadyPlate CT DG 18 Agar contact plates provide a ready-to-use method for environmental monitoring of surfaces in critical clean rooms and isolators. Dichloran Glycerol Medium is recommended for the isolation and enumeration of xerophilic molds from dried and semi-dried foods. The glycerol at 18% (w/w) lowers the water activity (aw) from 0.999 to 0.95 without causing any problem. Peptone provides carbon, nitrogen, vitamins, and minerals. Dextrose (Glucose) is a carbohydrate source. Phosphate buffers the medium. Magnesium sulfate provides divalent cations and sulfates. Dichloran is an antifungal agent added to the medium to reduce colony diameters of spreading fungi. Chloramphenicol is included to inhibit the growth of accompanying bacteria in the sample. Inhibiting the growth of bacteria and restriction of colony diameter of more-rapidly growing molds helps in the isolation of slow-growing fungi by preventing their overgrowth by more-rapidly growing species

Application

ReadyPlate CT Dichloran Glycerol Chloramphenicol Glucose Agar is used for the enumeration of yeast and molds from food and environmental samples with less water activity.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service