Skip to Content
Merck
All Photos(5)

Documents

1.32381

Millipore

Anaerocult A

To produce an anaerobic milieu in the anaerobic jar for the cultivation of obligatory and facultative anaerobes., suitable for microbiology

Synonym(s):

Anaerobic growth, Anaerobic incubation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104308
NACRES:
NB.14

Quality Level

form

solid

feature

Creates an oxygen free (anaerobic) environment and a CO2 atmosphere in a 2.5L anaerobic jar.

manufacturer/tradename

Anaerocult

packaging

pkg of 10 Anaerocult A

storage condition

(Store dry and tightly closed.

Keep in a well-ventilated place.)

color

white

application(s)

microbiology

compatibility

ANAEROTEST

storage temp.

15-25°C

General description

Anaerocult A is a reagent mixture used in microbiology to create an anaerobic environment to enable the growth of facultative and obligatory anaerobic microorganisms. Anaerocult A contains chemical components that bind the free oxygen in the atmosphere, producing carbon dioxide and establishing an anaerobic environment. The reaction is initiated by slightly moistening the reagent mixture, and incubation should be carried out in an Anaerobic jar or bag. For best results, Anaerocult A is used along with Anaerocult strips.

Application

The Anaerocult A system is used to produce an anaerobic milieu in the anaerobic jar for the incubation of up to 12 Petri dishes to cultivate obligatory and facultative anaerobes.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service