1.14764
Nitrate Test, photometric
photometric, 1.0-50.0 mg/L (NO3-N), 4-221 mg/L (NO3-), Spectroquant®
About This Item
Recommended Products
Product Name
Nitrate Cell Test, photometric, 1.0-50.0 mg/L (NO3-N), 4-221 mg/L (NO3-), Spectroquant®
product line
Spectroquant®
Quality Level
usage
sufficient for 25 tests
specific analyte(s)
nitrate
measuring range
1.0-50.0 mg/L (NO3-N)
4-221 mg/L (NO3-)
technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (2,6-Dimethylphenol)
storage temp.
15-25°C
General description
The method corresponds to ISO 23696-1, a norm standardizing the use of this "small-scale sealed tube method" for this important parameter. The method is also analogous to DIN 38405-9.
This Spectroquant® Nitrate Cell Test allows the accurate quantification of nitrate in water samples. Nitrate is an essential nutrient required for the growth of animals and plants. However, elevated levels of nitrate can be a health risk, in particular for pregnant women and infants. Therefore, nitrate levels in water sources are regulated in many countries.
Further fields of applications are the analysis of foods & beverages (e.g. whey) and of environmental and agricultural samples (e.g. soil).
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A step-by-step procedure for the photometric determination of Nitrate in effluents using Spectroquant® test kits and photometer for a precise and quantitative analysis.
Protocols
Learn about the photometric determination of nitrate ions in manure using Spectroquant® test kits and spectrophotometer.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service