Skip to Content
Merck
All Photos(1)

Key Documents

1.13175

Supelco

Spectromelt® A 1000

Li₂B₄O₇ + 0.07% LiBr, for X-ray fluorescence analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.21
form:
solid
solubility:
50 g/L

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

technique(s)

XRF: suitable

pH

9.1 (100 g/L)

mp

900 °C ((ca.1652 °F))

solubility

50 g/L

bulk density

500 kg/m3

anion traces

phosphate (PO43-): ≤20 ppm
silicate (as SiO2): ≤150 ppm
sulfate (SO42-): ≤50 ppm

cation traces

Ag: ≤5 ppm
Al: ≤10 ppm
Ba: ≤5 ppm
Ca: ≤20 ppm
Cd: ≤5 ppm
Co: ≤5 ppm
Cr: ≤5 ppm
Cu: ≤5 ppm
Fe: ≤10 ppm
Ga: ≤5 ppm
K: ≤10 ppm
Mg: ≤5 ppm
Mn: ≤5 ppm
Na: ≤20 ppm
Ni: ≤5 ppm
Pb: ≤5 ppm
Sr: ≤5 ppm
Zn: ≤5 ppm

storage temp.

2-30°C

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Phosphate (PO₄): ≤ 20 ppm
Silicate (SiO₂): ≤ 150 ppm
Sulfate (SO₄): ≤ 50 ppm
Ag (Silver): ≤ 5 ppm
Al (Aluminium): ≤ 10 ppm
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 20 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 10 ppm
Ga (Gallium): ≤ 5 ppm
K (Potassium): ≤ 10 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 5 ppm
Na (Sodium): ≤ 20 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Sr (Strontium): ≤ 5 ppm
Zn (Zinc): ≤ 5 ppm
Loss on ignition (800 °C): ≤ 1.0 %
Bulk density: about 60

Legal Information

SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service