1.10080
Manganese Test Strips, colorimetric
Mn 2-100 mg/L, MQuant®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116116
NACRES:
NA.24
Recommended Products
Product Name
Manganese Test, colorimetric, Mn 2-100 mg/L, MQuant®
description
semi-quantitative
Quality Level
product line
MQuant®
specific analyte(s)
manganese
reaction suitability
reagent type: oxidant
Mn measuring range
2-100 mg/L
detection method
colorimetric
storage temp.
15-25°C
Related Categories
General description
Method: Colorimetric, with test strips and reagent, 2 - 5 - 20 - 50 - 100 mg/L (Mn), MQuant®
The MQuant® Manganese test includes test strips and all necessary additional reagents and can be used for routine testing of water, industrial water and effluent. Manganese(II) ions are oxidized to manganese(IV) oxide, which transforms an organic redox indicator into a colored oxidation product. The manganese concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the manganese concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® test.
The MQuant® Manganese test includes test strips and all necessary additional reagents and can be used for routine testing of water, industrial water and effluent. Manganese(II) ions are oxidized to manganese(IV) oxide, which transforms an organic redox indicator into a colored oxidation product. The manganese concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the manganese concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® test.
Legal Information
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service