Skip to Content
Merck
All Photos(2)

Key Documents

1.09913

Supelco

Sodium hydroxide solution

1.000 l, for the preparation of dilute volumetric solutions or for direct use c(NaOH) = 5 mol/l (5 N) Combi-Titrisol®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

Product Name

Sodium hydroxide 1.000 l, for the preparation of dilute volumetric solutions or for direct use c(NaOH) = 5 mol/l (5 N) Combi-Titrisol

grade

reagent

Quality Level

vapor pressure

3 mmHg ( 37 °C)

product line

Titrisol®

form

liquid

pH

14 (20 °C in H2O)

density

1.185 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Silver Nanoparticles-Chitosan Nanocomposites: A Comparative Study Regarding Different Chemical Syntheses Procedures and Their Antibacterial Effect.: This study compares different chemical synthesis methods for creating silver nanoparticles-chitosan nanocomposites, focusing on their antibacterial properties. Sodium hydroxide is used as a reagent in these synthesis processes, demonstrating its role in developing effective antibacterial materials (Chicea et al., 2024).

  • Using Sparfloxacin-Capped Gold Nanoparticles to Modify a Screen-Printed Carbon Electrode Sensor for Ethanol Determination.: This research develops a sensor for ethanol detection using gold nanoparticles capped with sparfloxacin. Sodium hydroxide is involved in the modification process of the screen-printed carbon electrode, highlighting its utility in sensor development (Magesh et al., 2023).

  • Sequence Mining Yields 18 Phloretin C-Glycosyltransferases from Plants for the Efficient Biocatalytic Synthesis of Nothofagin and Phloretin-di-C-glycoside.: The study identifies new enzymes for synthesizing glycosylated compounds from plants. Sodium hydroxide is used in the biocatalytic processes, showcasing its importance in biochemical synthesis (Putkaradze et al., 2023).

  • Effect of Calcination Temperature and Time on the Synthesis of Iron Oxide Nanoparticles: Green vs. Chemical Method.: This research examines the synthesis of iron oxide nanoparticles, comparing green and chemical methods. Sodium hydroxide is used in the chemical synthesis route, demonstrating its significance in nanoparticle production (Abdullah et al., 2023).

  • The Use of NaOH Solutions for Fouling Control in a Membrane Bioreactor: A Feasibility Study.: This study explores the application of sodium hydroxide solutions for controlling fouling in membrane bioreactors, highlighting its effectiveness in maintaining bioreactor efficiency (Tomczak et al., 2021).

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service