Skip to Content
Merck
All Photos(1)

Key Documents

1.09609

Supelco

n-Tridecane

reference substance for gas chromatography

Synonym(s):

n-Tridecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CH3
CAS Number:
Molecular Weight:
184.36
MDL number:
UNSPSC Code:
12352001
EC Index Number:
211-093-4

vapor pressure

<0.5 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

>204 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

0.6-6.5 % (v/v)

technique(s)

gas chromatography (GC): suitable

bp

235 °C/1013 hPa

mp

-5 °C

transition temp

flash point 79 °C

density

0.757 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C13H28/c1-3-5-7-9-11-13-12-10-8-6-4-2/h3-13H2,1-2H3

InChI key

IIYFAKIEWZDVMP-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99.3 % (a/a)
Identity (IR-spectrum): conforms

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service