Skip to Content
Merck
All Photos(2)

Documents

1.08382

Millipore

Tris Base

solid, ACS reagent, reag. Ph. Eur., GR for analysis buffer substance

Synonym(s):

Tris Base, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
MDL number:
UNSPSC Code:
12161703
EC Index Number:
201-064-4
NACRES:
NA.05

product name

Tris(hydroxymethyl)aminomethane, GR for analysis buffer substance ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

0.0267 hPa ( 20 °C)

form

solid

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

pH

10.2-10.6 (20 °C, 6 g/L in H2O)

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)
219-220 °C/13.3 hPa

mp

167-172 °C (lit.)

solubility

678 g/L

density

1.35 g/cm3 at 23 °C

bulk density

840 kg/m3

storage temp.

15-25°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biological buffer substance
Tris is widely used as a biological buffer substance in the pH range of 7-9. For example it is used for the freezing of proteins. The grade conforms precisely with the Reag. Ph. Eur. requirements.
GR for analysis buffer substance ACS,Reag. Ph Eur

Application

Tris(hydroxymethyl)aminomethane GR for analysis buffer substance ACS,Reag. Ph Eur. CAS 77-86-1, EC 201-064-4, chemical formula H₂NC(CH₂OH)₃.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service