1.07729
Silica gel 60 (less than 0.063 mm)
for column chromatography
Synonym(s):
Silica, Silicon dioxide
About This Item
Recommended Products
Quality Level
form
solid
matrix active group
Irregular-Shaped Silica Gel
pH
7 (20 °C, 100 g/L in H2O, slurry)
bp
2230 °C (lit.)
mp
>1600 °C (lit.)
bulk density
200‑1430 kg/m3
separation technique
normal phase
storage temp.
no temp limit
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- High adsorptive capacity
- High chemical inertness and non-toxicity.
- Protective moisture-sensitive substances during storage and transport or for maintaining the dryness of anhydrous solvents.
- Granulates in different sizes or beads
Packaging
Analysis Note
Fe (Iron): ≤ 0.02 %
Loss on drying: ≤ 7.0 %
Particle size: 95 % < 0,063 mm
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service