Skip to Content
Merck
All Photos(2)

Key Documents

1.07559

Supelco

Extran® AP 21

greener alternative

liquid, acidic, concentrate (contains phosphoric acid)

Synonym(s):

TM=["EXTRAN"] AP 21 cleaning agent; TM=["EXTRAN"] AP 21 detergent

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$144.30
250 MG
$206.70
500 MG
$325.00
1 G
$416.00
2.5 G
$655.20
5 G
$1,053.00
10 G
$1,677.00

Vendor SKU: EN300-105082-100MG

$144.30


Ships in 7 days - from Aldrich Partner.

Request a Bulk Order

Select a Size

Change View
100 MG
$144.30
250 MG
$206.70
500 MG
$325.00
1 G
$416.00
2.5 G
$655.20
5 G
$1,053.00
10 G
$1,677.00

About This Item

UNSPSC Code:
41103206
NACRES:
NA.21

Vendor SKU: EN300-105082-100MG

$144.30


Ships in 7 days - from Aldrich Partner.

Request a Bulk Order

Quality Level

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

53.0-57.0% (acidimetric)

pH

2.0 (20 °C, 2 g/L in H2O)

density

1.37 g/cm3 at 20 °C

greener alternative category

storage temp.

no temp limit

General description

This product is identical to EX1003 with the same quality and specification. Some pack sizes have changed to provide a globally harmonized offering.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is environmentally friendly having bio-degradable active ingredients. Click here for more information.

Linkage

Replaces: EX1003-4

Analysis Note

Assay (acidimetric): 53.0 - 57.0 %

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Select a Product Number

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service