Skip to Content
Merck
All Photos(1)

Documents

1.07227

Supelco

Phenolphthalein solution

1% in ethanol, indicator pH 8.2 - 9.8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171502
NACRES:
NA.21

product name

Phenolphthalein solution 1% in ethanol, indicator pH 8.2 - 9.8

Agency

suitable for SM 2310
suitable for SM 2320

Quality Level

form

liquid

autoignition temp.

425 °C

transition temp

flash point 23 °C

density

0.89 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Application test: passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service