Skip to Content
Merck
All Photos(1)

Key Documents

1.04063

Supelco

Glycerol solution

Synonym(s):

Glycerol solution, Glycerin, 1,2,3-Propanetriol, Trihydroxylpropane, Protol, 1,2,3-Propanetriol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH · aq
CAS Number:
Molecular Weight:
92.09 (anhydrous basis)
Beilstein:
635685
MDL number:
UNSPSC Code:
50161901
bp:
182 °C/20 mmHg (lit.)

form

liquid

Quality Level

pH

~5 (20 °C, 100 g/L)

bp

182 °C/20 mmHg (lit.)

mp

-10 °C

density

1.252 g/mL at 25 °C (lit.)

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service