Skip to Content
Merck
All Photos(1)

Key Documents

1.02529

Supelco

Cobalt(II) acetate tetrahydrate

for analysis EMSURE® ACS

Synonym(s):

Cobalt(II) acetate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Co · 4 H2O
CAS Number:
Molecular Weight:
249.08
MDL number:
UNSPSC Code:
12352103
EC Index Number:
200-755-8

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

99.0-101.0% (complexometric)

form

solid

impurities

≤0.01% Insoluble matter

pH

7.2 (20 °C, 50 g/L in H2O)

mp

140 °C (Elimination of water of crystallisation)

solubility

348 g/L

density

1.70 g/cm3 at 20 °C

bulk density

850 kg/m3

anion traces

chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.01%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
N: ≤0.002%
Na: ≤0.01%
Ni: ≤0.02%
Pb: ≤0.0005%
Zn: ≤0.001%

storage temp.

2-30°C

InChI

1S/2C2H4O2.Co.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

ZBYYWKJVSFHYJL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (complexometric): 99.0 - 101.0 %
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water): 6.0 - 8.0
Chloride (Cl): ≤ 0.001 %
Nitrate (NO₃): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.002 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.005 %
Na (Sodium): ≤ 0.01 %
Ni (Nickel): ≤ 0.02 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service