Skip to Content
Merck
All Photos(2)

Key Documents

1.00849

Supelco

Toluene

for gas chromatography MS Suprasolv®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-625-9
NACRES:
NA.03
technique(s):
GC/MS: suitable
gas chromatography (GC): suitable
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

vapor density

3.2 (vs air)

Quality Level

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

GC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.03% Water

evapn. residue

≤3.0 mg/L

color

APHA: ≤10

refractive index

n/D 1.496 (lit.)

kinematic viscosity

0.7 cSt(20 °C)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

transition temp

flash point 4 °C

solubility

0.52 g/L

density

0.865 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Application


  • Analytical Solution for The Toluene and Mixture of Toluene and n-Propanol in The Biofilm Using ADM.: This study provides an analytical solution for the diffusion and reaction of toluene and its mixtures with n-propanol in biofilms using the Adomian Decomposition Method (ADM). The findings highlight the efficiency of ADM in solving complex diffusion-reaction problems in environmental engineering applications (Valli et al., 2020).

  • Benzene, toluene, ethylbenzene, and xylene: Current analytical techniques and approaches for biological monitoring.: This comprehensive review discusses various analytical techniques and biological monitoring approaches for assessing human exposure to toluene and other BTEX compounds. It evaluates the effectiveness of different sample preparation methods and analytical techniques, emphasizing the importance of reliable biomarkers for occupational exposure (Soleimani, 2020).

  • Employing UV/peroxydisulphate (PDS) activated by ferrous ion for the removal of toluene in aqueous environment: electrical consumption and kinetic study.: This research investigates the removal of toluene from aqueous environments using UV/peroxydisulphate activated by ferrous ions. The study provides insights into the kinetic parameters and electrical consumption of the process, demonstrating its potential for environmental remediation applications (Shokri, 2020).

  • Investigation of some effective factors on urinary metabolites in biological monitoring of benzene, toluene, and xylene compounds.: This paper explores the factors influencing urinary metabolites in the biological monitoring of exposure to benzene, toluene, and xylene (BTX) compounds. It highlights the importance of understanding these factors for accurate assessment of occupational exposure to BTX compounds (Taheri et al., 2022).

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service