Skip to Content
Merck
All Photos(1)

Key Documents

07-472

Sigma-Aldrich

Anti-PTPα Antibody

Upstate®, from rabbit

Synonym(s):

Anti-HEPTP, Anti-HPTPA, Anti-HPTPalpha, Anti-LRP, Anti-PTPA, Anti-PTPRL2, Anti-R-PTP-alpha, Anti-RPTPA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PTPRA(5786)
mouse ... Ptpra(19262)
rat ... Ptpra(25167)

Specificity

PTP alpha isoform

Immunogen

N-terminal GST-tagged fusion protein corresponding to residues 505-793 of human PTP alpha (complete D2 catalytic domain).

Application

Anti-PTPα Antibody detects level of PTPα & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

Routinely evaluated by immunoblot in in NIH 3T3 cell lysates.

Target description

130kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
ImmunoAffinity Purified

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An RPTP?/Src family kinase/Rap1 signaling module recruits myosin IIB to support contractile tension at apical E-cadherin junctions.
Gomez, GA; McLachlan, RW; Wu, SK; Caldwell, BJ; Moussa, E; Verma, S; Bastiani, M; Priya et al.
Molecular Biology of the Cell null
Two mechanisms activate PTPalpha during mitosis.
Zheng, X M and Shalloway, D
The Embo Journal, 20, 6037-6049 (2001)
A Petrone et al.
Journal of cell science, 113 ( Pt 13), 2345-2354 (2000-06-15)
Transmembrane (receptor) tyrosine phosphatases are intimately involved in responses to cell-cell and cell-matrix contact. Several important issues regarding the targets and regulation of this protein family are now emerging. For example, these phosphatases exhibit complex interactions with signaling pathways involving
Chan Zhou et al.
Genome medicine, 8(1), 31-31 (2016-03-24)
Hepatic fibrosis is the underlying cause of cirrhosis and liver failure in nearly every form of chronic liver disease, and hepatic stellate cells (HSCs) are the primary cell type responsible for fibrosis. Long noncoding RNAs (lncRNAs) are increasingly recognized as
Eliot T McKinley et al.
PloS one, 8(11), e80207-e80207 (2013-11-22)
Sensitive and specific biomarkers of protein kinase inhibition can be leveraged to accelerate drug development studies in oncology by associating early molecular responses with target inhibition. In this study, we utilized unbiased shotgun phosphotyrosine (pY) proteomics to discover novel biomarkers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service