Skip to Content
Merck
All Photos(1)

Key Documents

860924P

Avanti

N-24:0(2R-OH) Phytosphingosine

Avanti Research - A Croda Brand 860924P, powder

Synonym(s):

N-(2′-(R)-hydroxylignoceroyl)-phytosphingosine (Saccharomyces Cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H85NO5
CAS Number:
Molecular Weight:
684.13
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860924P-10mg)
pkg of 1 × 5 mg (860924P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860924P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@]([H])(CCCCCCCCCCCCCC)[C@](O)([H])[C@@]([H])(NC([C@H](O)CCCCCCCCCCCCCCCCCCCCCC)=O)CO

InChI

1S/C42H85NO5/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-40(46)42(48)43-38(37-44)41(47)39(45)35-33-31-29-27-25-16-14-12-10-8-6-4-2/h38-41,44-47H,3-37H2,1-2H3,(H,43,48)

InChI key

ZFUXWVVVWGWGPQ-UHFFFAOYSA-N

General description

N-24:0(2R-OH) Phytosphingosine, also called as N-(2′-(R)-hydroxylignoceroyl)-phytosphingosine, is a sphingolipid that belongs to the group of phytoceramides. Phytosphingosine is a sphingoid base with hydroxyl group at the C4 position. It is highly found in plants and fungi. In addition, phytosphingosine is also present in mammalian epidermis, including the stratum corneum.

Application

N-24:0(2R-OH) Phytosphingosine, also called as N-(2′-(R)-hydroxylignoceroyl)-phytosphingosine may be used to study its effects in different hybrids of Zea mays.

Biochem/physiol Actions

Phytosphingosine and its derivatives play a vital role in the body′s natural defense system. These lipids prevent dehydration of skin, control epidermal cell growth, differentiation and apoptosis. Phytosphingosine also exhibits anti-bactericidal and anti-inflammatory properties.

Packaging

5 mL Amber Glass Screw Cap Vial (860924P-10mg)
5 mL Amber Glass Screw Cap Vial (860924P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Giorni et al.
Toxins, 7(9), 3657-3670 (2015-09-18)
Lipid molecules are increasingly recognized as signals exchanged by organisms interacting in pathogenic and/or symbiotic ways. Some classes of lipids actively determine the fate of the interactions. Host cuticle/cell wall/membrane components such as sphingolipids and oxylipins may contribute to determining
Chiara Mencarelli et al.
Cellular and molecular life sciences : CMLS, 70(2), 181-203 (2012-06-26)
Ceramide, the precursor of all complex sphingolipids, is a potent signaling molecule that mediates key events of cellular pathophysiology. In the nervous system, the sphingolipid metabolism has an important impact. Neurons are polarized cells and their normal functions, such as
Byung-Hak Kim et al.
The Journal of investigative dermatology, 134(4), 1023-1032 (2013-11-02)
Phytosphingosine is abundant in plants and fungi and is found in mammalian epidermis, including the stratum corneum. Phytosphingosine and its derivatives N-acetyl phytosphingosine and tetraacetyl phytosphingosine are part of the natural defense system of the body. However, these molecules exhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service