Skip to Content
Merck
All Photos(2)

Key Documents

857132P

Avanti

14:0 Hemi BMP (S,R)

sn-(3-myristoyl-2-hydroxy)-glycerol-1-phospho-sn-3′-(1′,2′-dimyristoyl)-glycerol (ammonium salt), powder

Synonym(s):

110731

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H96NO11P
CAS Number:
Molecular Weight:
894.25
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (857132P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857132P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(OC[C@@]([H])(O)COC(CCCCCCCCCCCCC)=O)([O-])=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C48H93O11P.H3N/c1-4-7-10-13-16-19-22-25-28-31-34-37-46(50)55-40-44(49)41-57-60(53,54)58-43-45(59-48(52)39-36-33-30-27-24-21-18-15-12-9-6-3)42-56-47(51)38-35-32-29-26-23-20-17-14-11-8-5-2;/h44-45,49H,4-43H2,1-3H3,(H,53,54);1H3/t44-,45+;/m0./s1

InChI key

LRUIYSPDUHVXNZ-OOWSDCSBSA-N

General description

Hemi-bis(monoacylglycero)phosphate (hemi-BMP) consists of a glycerophosphoglycerol backbone wherein the three fatty acyl chains are esterified to the glycerol hydroxyls.

Packaging

5 mL Amber Glass Screw Cap Vial (857132P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hemi-bis (monoacylglycero) phosphate composition and biosynthesis in Escherichia coli
Garrett TA and Rose RL
Faseb Journal (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service