Skip to Content
Merck
All Photos(2)

Key Documents

855779P

Avanti

22:0 Lyso PC

1-behenoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-docosanoyl-sn-glycero-3-phosphocholine; PC(22:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H62NO7P
CAS Number:
Molecular Weight:
579.79
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855779P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855779P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCCCCCCCCC)=O

InChI

1S/C30H62NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-30(33)36-27-29(32)28-38-39(34,35)37-26-25-31(2,3)4/h29,32H,5-28H2,1-4H3/t29-/m1/s1

InChI key

UIINDYGXBHJQHX-GDLZYMKVSA-N

Application

22:0 Lyso PC may be used as a standard to screen X-linked adrenoleukodystrophy (X-ALD) in newborns using positive ion electrospray (ESI) combined liquid chromatography-tandem spectrometric (LC-MS/MS) technique.

Packaging

5 mL Amber Glass Screw Cap Vial (855779P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Newborn screening for X-linked adrenoleukodystrophy (X-ALD): validation of a combined liquid chromatography-tandem mass spectrometric (LC-MS/MS) method
Hubbard WC, et al.
Molecular Genetics and Metabolism, 97(3), 212-220 (2009)
Walter C Hubbard et al.
Molecular genetics and metabolism, 89(1-2), 185-187 (2006-07-11)
Utilizing combined liquid chromatography-tandem mass spectrometry (LC-MS/MS) as the analytical method, we have demonstrated a ten to sixtyfold excess of lysophosphatidyl choline containing hexacosanoic acid (26:0) in dried blood spots on a filter paper matrix from 25 male patients with
Francesca Saitta et al.
Colloids and surfaces. B, Biointerfaces, 186, 110715-110715 (2019-12-17)
A fifteen-components model membrane that reflected the 80 % of phospholipids present in Insulin Secretory Granules was obtained and thermodynamic exploitation was performed, through micro-DSC, in order to assess the synergic contributions to the stability of a mixed complex system
Kelsey B Law et al.
Autophagy, 13(5), 868-884 (2017-05-20)
Peroxisome biogenesis disorders (PBDs) are metabolic disorders caused by the loss of peroxisomes. The majority of PBDs result from mutation in one of 3 genes that encode for the peroxisomal AAA ATPase complex (AAA-complex) required for cycling PEX5 for peroxisomal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service