Skip to Content
Merck
All Photos(1)

Key Documents

810105P

Avanti

palmitic acid - NBD

Avanti Research - A Croda Brand

Synonym(s):

[16-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino] palmitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34N4O5
CAS Number:
Molecular Weight:
434.53
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810105P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

16-NBD palmitic acid (16-NBD) is a fluorescent lipid. NBD fluorophore (7-nitrobenzenz- 2-oxa-1,3-diazol-4-yl) elicits visible range fluorescence.

Application

Palmitic acid - NBD may be used as fluorophore in lipid formulations for fluorescent labeling of nanoparticle (NP) to trace localization in skin sections.

Biochem/physiol Actions

16-NBD palmitic acid has been used in nanoparticle localization studies. NBD fluorophore (7-nitrobenzenz- 2-oxa-1,3-diazol-4-yl) is used in generating fluorescent phospholipids and its conjugate in lipids is useful as a quencher in membrane phase separation studies.

Packaging

5 mL Amber Glass Screw Cap Vial (810105P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Altered membrane lipid dynamics and chemoprevention by non-steroidal anti inflammatory drugs during colon carcinogenesis
Kanwar SS, et al.
Nutricion Hospitalaria : Organo Oficial de la Sociedad Espanola de Nutricion Parenteral y Enteral, 26(5), 1141-1154 (2011)
Development and in vitro evaluation of lipid nanoparticle-based dressings for topical treatment of chronic wounds
Gainza G, et al.
International Journal of Pharmaceutics, 490(1-2), 404-411 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service