Skip to Content
Merck
All Photos(1)

Documents

700166P

Avanti

Sitoindoside II

6-O-oleoyl-sitosteryl-β-D-glucose, powder

Synonym(s):

3-O-[(6′-O-oleoyl)-β-D-glucopyranosyl] sitosterol; 18:1 sitosteryl glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H92O7
CAS Number:
Molecular Weight:
841.29
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700166P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700166P

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C53H92O7/c1-8-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-47(54)58-36-46-48(55)49(56)50(57)51(60-46)59-41-31-33-52(6)40(35-41)27-28-42-44-30-29-43(53(44,7)34-32-45(42)52)38(5)25-26-39(9-2)37(3)4/h16-17,27,37-39,41-46,48-51,55-57H,8-15,18-26,28-36H2,1-7H3/b17-16-/t38-,39+,41+,42+,43-,44+,45+,46-,48-,49+,50-,51-,52+,53-/m1/s1

InChI key

MEEONOMPSMYAQO-JXJRCNQOSA-N

General description

Sitoindoside-II is an acyl steryl glycoside present in M. paradisiaca (banana) fruits. It is also isolated from the chicory plant, Cichorium intybus L. Sitoindoside-II is a fatty acid ester of steroidal glucopyranoside.

Packaging

5 mL Amber Glass Screw Cap Vial (700166P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R O Vilhena et al.
Journal of ethnopharmacology, 254, 112666-112666 (2020-02-23)
The Musa x paradisiaca L. inflorescence, known as banana blossom or banana heart, is used in traditional medicine for the treatment of diabetes mellitus. The aim of the study was to investigate the antidiabetic activity of aqueous extracts and fractions
Dinara Satmbekova et al.
Natural product research, 32(11), 1343-1347 (2017-06-21)
Cichorium intybus L. (Asteraceae family) is a world-wide grown plant known as chicory. In traditional medicine, this plant is used as diuretic, anti-inflammatory, digestive, cardiotonic and liver tonic. Chromatographic purification of the supercritical fluid extract of aerial parts of C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service