Skip to Content
Merck
All Photos(1)

Key Documents

330714W

Avanti

Deuterated Psychosine Quantitative Mass Spec Standard

Avanti Research - A Croda Brand

Synonym(s):

Galactosyl(β) sphingosine-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42D5NO7
Molecular Weight:
466.66
UNSPSC Code:
41116107
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (330714W-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

0.45 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCC([2H])(C([2H])([2H])[2H])[2H])(O)[C@](N)([H])CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI key

HHJTWTPUPVQKNA-LTNFHSIGSA-N

Packaging

2 mL Amber Glass Sealed Ampule (330714W-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam J Guenzel et al.
Genetics in medicine : official journal of the American College of Medical Genetics (2020-02-25)
Newborn screening (NBS) for Krabbe disease (KD) is performed by measurement of galactocerebrosidase (GALC) activity as the primary test. This revealed that GALC activity has poor specificity for KD. Psychosine (PSY) was proposed as a disease marker useful to reduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service