Key Documents
Z682403
Ace glass steam tube, for condenser "C", "CR", "A" and "V" assemblies
fits Büchi® models R-200/R-205 and R-114/R-144
Synonym(s):
40610
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
manufacturer/tradename
Büchi equivalent, 40610
Related Categories
General description
The glass steam tube accessory fits the Büchi condenser assemblies, "C", "CR", "A" and "V", and works with Büchi Rotavapor models R-200/R-205 and R-114/R-144.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)