Z563293
Plas-Labs multi-station Nitrogen dry-box glove box
AC/DC input 240 V AC, Euro plug
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
Recommended Products
grade
CE Compliant
manufacturer/tradename
Plas-Labs 850-NBB/EXP
H × W × D
28 in. × 60 in. × 38 in., inside dimensions
42 in. × 76 in. × 47 in., outside dimensions
weight
300 lb
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Clear, one piece unit with molded base section
- Adjustable SS clamps compensate for wear
- Two pairs of gloves ports for two or more operators working intandem.
- 8 in. O.D. ports with ambidextrous Hypalon® gloves with SS O-rings
- Transparent transfer chamber (12 L × 11 in. i.d.) with adjustable vacuum gauge
- Gas-drying train with DesiSphere desiccating canisters, self-sealing quick-disconnects enable change of drying train without disturbing internal atmosphere
- Two vacuum/pressure pumps - one for transfer chamber, one for drying train to speed purging.
- Fluorescent light system with illuminated controls
- Self-sealing quick disconnects enable the operator to change the drying train without disturbing the internal atmosphere.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service