Z154466
ACE PTFE-coated glass shaft
diam. × L 10 mm × 44 cm, with button
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
for flask size 250 ‑ 5,000 mL
manufacturer/tradename
Ace Glass 807005
diam. × L
10 mm × 44 cm
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service