Key Documents
Z108286
Tempo-Shield heat-resistant gloves
L 18 in.
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
Kevlar® (Asbestos-free heat-resistant gloves with wool liner)
size
one size fits all
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pkg of 1 pair
manufacturer/tradename
Sigma-Aldrich
L
18 in.
General description
Asbestos-free heat-resistant gloves with wool liner provide strength and heat protection up to 500 °F. Made from a blend of aluminized and synthetic fibers. Ambidextrous.
Legal Information
Kevlar is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)