Key Documents
WHAMUPG2MCWT8
Whatman® Mini-UniPrep® G2 hand compressor
8 position tray, pack of 1
Synonym(s):
hand compressor, mini-uniprep G2, syringeless filter, whatman filter
About This Item
UNSPSC Code:
40161807
NACRES:
NB.24
packaging
pack of 1
manufacturer/tradename
Cytiva
Whatman Article No. 29036891 (US reference)
Whatman MUPG2MCWT8
General description
Mini-UniPrep G2 Hand Compressor is for use with Mini-UniPrep G2 Syringeless filters.
Features and Benefits
  • A multicompressor streamlines syringeless filtration – preload in advance with an extra tray
  • Designed for Cytiva′s Mini-UniPrepG2, which combines a borosilicate glass vial, plunger and attached filter membrane, and septum with cap
Legal Information
Mini-UniPrep is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva