WHA10534612
Whatman® protein saver cards
903 Protein saver card (US), pkg of 100 ea
Synonym(s):
Whatman fta, Z761575, dbs cards, pcr blood
About This Item
Recommended Products
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 10534612
Whatman Article No. 28416778 (US reference)
Looking for similar products? Visit Product Comparison Guide
General description
- Storage: each circle holds 75-80 µL of sample
- Strict quality control and ISO9001 manufacturing standards: ensures high quality, reproducibility, and purity
Features and Benefits
- The Proteinsaver card has a wraparound cover with spaces for name and date of collection.
- The card fits into our Whatman foil barrier resealable bags for storage.
- Imprinted with the Universal biohazard symbol in accordance with USP regulations.
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
In this article we provide a complete workflow of quantification and identification of acylcarnitines in dried blood spots (DBS) using stable isotope-labeled (SIL) acylcarnitines CRMs by FIA-MS/MS.
Protocols
A number of options are available for sample collection, transport, archiving, and DNA purification.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service