GF59182612
Polyimide
(PI), laminate, thickness 0.1 mm, size 100 × 100 mm, polymer thickness 0.05mm
Synonym(s):
IM301536
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Backing Material: 0.035mm Copper
form
film
manufacturer/tradename
Goodfellow 591-826-12
size
100 × 100 mm
thickness
0.1 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service