Skip to Content
Merck
All Photos(2)

Documents

GF45554799

Lithium

rod, 12.7 mm diameter, length 165 mm, purity 99%

Synonym(s):

LI007915

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li
CAS Number:
Molecular Weight:
6.94
MDL number:
UNSPSC Code:
12141803
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

rod

manufacturer/tradename

Goodfellow 455-547-99

resistivity

9.446 μΩ-cm, 20°C

L

165 mm

diameter

12.7 mm

bp

1342 °C (lit.)

mp

180 °C (lit.)

density

0.534 g/mL at 25 °C (lit.)

SMILES string

[Li]

InChI

1S/Li

InChI key

WHXSMMKQMYFTQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service