DWKCPB0010P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations
class B, capacity 10 mL , with +/-0.2mL tolerance
Synonym(s):
CPB0010P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121805
NACRES:
NA.54
Recommended Products
feature
class B
graduated
packaging
pkg of 10 ea
manufacturer/tradename
Azlon CPB0010P
capacity
10 mL , with +/-0.2mL tolerance
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Suitable for the measurement and dispensing of liquids in the laboratory
Features and Benefits
Conform to ISO 6706 with anti-suction designto prevent base sticking to wet surface
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service